Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LHY
Entrez Gene ID
839341
Gene Name
Homeodomain-like superfamily protein
Gene Aliases
AT1G01060, LATE ELONGATED HYPOCOTYL, LATE ELONGATED HYPOCOTYL 1, LHY1, T25K16.6, T25K16_6
Chromosome Location
Chr.1: 33365 - 37871 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035847.1 NP_001030924.1 8 - 9 1962 91
NM_001083968.1 NP_001077437.1 8 - 9 1982 91
NM_001197953.1 NP_001184882.1 8 - 9 2364 91
NM_001331246.1 NP_001320628.1 8 - 9 2364 91
NM_001331247.1 NP_001320627.1 7 - 8 2245 91
NM_001331248.1 NP_001320629.1 6 - 7 1774 91
NM_099988.4 NP_171614.1 8 - 9 2045 91
NM_179237.1 NP_849568.1 7 - 8 2155 91
GenBank mRNA AK316728.1 - 7 - 8 2155 91

View Full Product Details