Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPSRP54
Entrez Gene ID
830273
Gene Name
chloroplast signal recognition particle 54 kDa subunit
Gene Aliases
54 CHLOROPLAST PROTEIN, 54CP, AT5G03940, F8F6.150, F8F6_150, FFC, FIFTY-FOUR CHLOROPLAST HOMOLOGUE, SIGNAL RECOGNITION PARTICLE 54 KDA SUBUNIT, SIGNAL RECOGNITION PARTICLE 54 KDA SUBUNIT CHLOROPLAST PROTEIN, SRP54CP, chloroplast signal recognition particle 54 kDa subunit
Chromosome Location
Chr.5: 1060106 - 1063425 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120476.5 NP_196014.1 13 - 14 1761 63
GenBank mRNA AY050999.1 - 13 - 14 1657 63
BT000976.1 - 13 - 14 1593 63
Z21970.1 - - 1616 63

View Full Product Details