Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APC10
Entrez Gene ID
816345
Gene Name
anaphase promoting complex 10
Gene Aliases
AT2G18290, AtAPC10, EMB2783, EMBRYO DEFECTIVE 2783, T30D6.20, T30D6_20, anaphase promoting complex 10
Chromosome Location
Chr.2: 7948306 - 7950329 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127387.3 NP_565433.1 2 - 3 472 141
GenBank mRNA AK229538.1 - 2 - 3 347 141
BT024912.1 - 2 - 3 239 141

View Full Product Details