Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XIG
Entrez Gene ID
816548
Gene Name
myosin-like protein XIG
Gene Aliases
AT2G20290, ATXIG, F11A3.16, F11A3_16, MYOSIN XI G, myosin-like protein XIG
Chromosome Location
Chr.2: 8742885 - 8752315 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335672.1 NP_001325368.1 24 - 25 3157 134
NM_001335673.1 NP_001325369.1 24 - 25 3157 134
NM_001335674.1 NP_001325367.1 24 - 25 3164 134
NM_127588.3 NP_179619.2 24 - 25 2966 134

View Full Product Details