Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RR24
Entrez Gene ID
832707
Gene Name
response regulator 24
Gene Aliases
ARR24, AT5G26594, response regulator 24
Chromosome Location
Chr.5: 9269282 - 9270254 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180533.1 NP_850864.1 1 - 2 238 101

View Full Product Details