Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- G18E
- Entrez Gene ID
-
830397
- Gene Name
- autophagy-related protein 18E
- Gene Aliases
-
ARABIDOPSIS THALIANA HOMOLOG OF YEAST AUTOPHAGY 18E, AT5G05150, ATATG18E, ATG18E, K2A11.2, K2A11_2, autophagy-related gene 18E
- Chromosome Location
-
Chr.5: 1524699 - 1526199 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120597.2 |
NP_196134.1 |
1 - 2 |
731 |
86 |
GenBank mRNA |
DQ056670.1 |
- |
1 - 2 |
731 |
86 |
View Full Product Details