Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNAP29
Entrez Gene ID
830681
Gene Name
synaptosomal-associated protein SNAP25-like 29
Gene Aliases
ARABIDOPSIS THALIANA SYNAPTOSOMAL-ASSOCIATED PROTEIN SNAP25-LIKE 29, AT5G07880, ATSNAP29, F13G24.80, F13G24_80, synaptosomal-associated protein SNAP25-like 29
Chromosome Location
Chr.5: 2515219 - 2517066 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342964.1 NP_001318503.1 5 - 6 808 86
NM_120870.2 NP_196405.1 4 - 5 716 86
GenBank mRNA DQ056673.1 - 4 - 5 653 86

View Full Product Details