Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUB22
Entrez Gene ID
824410
Gene Name
plant U-box 22
Gene Aliases
AT3G52450, AtPUB22, plant U-box 22
Chromosome Location
Chr.3: 19440617 - 19442427 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115105.4 NP_190813.1 1 - 1 1201 105
GenBank mRNA AK228993.1 - 1 - 1 1157 105
BT015763.1 - 1 - 1 1121 105
BT020203.1 - 1 - 1 1024 105

View Full Product Details