Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIGC
Entrez Gene ID
824559
Gene Name
RNApolymerase sigma-subunit C
Gene Aliases
AT3G53920, RNA POLYMERASE SIGMA-SUBUNIT, RNApolymerase sigma-subunit C, SIG3, SIGMA FACTOR 3
Chromosome Location
Chr.3: 19960789 - 19963980 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339642.1 NP_001326036.1 6 - 7 1248 67
NM_115252.3 NP_190960.1 6 - 7 1248 67
GenBank mRNA AF015544.1 - 6 - 7 1097 67
BT030362.1 - 6 - 7 1088 67
D89994.1 - 6 - 7 1217 67

View Full Product Details