Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YSL8
Entrez Gene ID
841257
Gene Name
YELLOW STRIPE like 8
Gene Aliases
AT1G48370, F11A17.8, F11A17_8, YELLOW STRIPE like 8
Chromosome Location
Chr.1: 17874436 - 17877462 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103733.4 NP_564525.1 1 - 2 585 109
GenBank mRNA AY039600.1 - 1 - 2 505 109
BT010472.1 - 1 - 2 461 109

View Full Product Details