Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- UBA1
- Entrez Gene ID
-
817562
- Gene Name
- ubiquitin-activating enzyme 1
- Gene Aliases
-
AT2G30110, ATUBA1, MODIFIER OF SNC1 5, MOS5, T27E13.15, T27E13_15, ubiquitin-activating enzyme 1
- Chromosome Location
-
Chr.2: 12852329 - 12857727 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128566.4 |
NP_565693.1 |
5 - 6 |
2443 |
66 |
View Full Product Details