Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- P5CR
- Entrez Gene ID
-
831332
- Gene Name
- pyrroline-5- carboxylate (P5C) reductase
- Gene Aliases
-
AT-P5C1, AT-P5R, AT5G14800, EMB2772, EMBRYO DEFECTIVE 2772, PYRROLINE-5- CARBOXYLATE (P5C) REDUCTASE, T9L3.100, T9L3_100, pyrroline-5- carboxylate (P5C) reductase
- Chromosome Location
-
Chr.5: 4785960 - 4787996 on Build TAIR10.1
- Gene Symbol
- AT4G18260
- Entrez Gene ID
-
827552
- Gene Name
- Cytochrome b561/ferric reductase transmembrane protein family
- Gene Aliases
-
AT4G18260, T9A21.110, T9A21_110
- Chromosome Location
-
Chr.4: 10095658 - 10097337 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK228989.1 |
- |
- |
1385 |
68 |
View Full Product Details