Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCBP
Entrez Gene ID
831929
Gene Name
Putative cap-binding protein
Gene Aliases
AT5G18110, MRG7.7, MRG7_7, novel cap-binding protein
Chromosome Location
Chr.5: 5988744 - 5990681 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121816.2 NP_197312.1 3 - 4 430 95
GenBank mRNA AY092964.1 - 3 - 4 430 95
AY093208.1 - 3 - 4 426 95
AY114562.1 - 3 - 4 337 95

View Full Product Details