Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XIF
Entrez Gene ID
817748
Gene Name
myosin-like protein XIF
Gene Aliases
AT2G31900, ATMYO5, ATXIF, F20M17.6, F20M17_6, MYOSIN 5, MYOSIN XI F, myosin-like protein XIF
Chromosome Location
Chr.2: 13560370 - 13570469 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336351.1 NP_001318329.1 35 - 36 4879 103
NM_001336352.1 NP_001323600.1 33 - 34 4169 103
NM_001336353.1 NP_001323601.1 28 - 29 3395 103
NM_001336354.1 NP_001323599.1 31 - 32 3960 103
NM_128748.3 NP_180749.2 33 - 34 4166 103

View Full Product Details