Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XIF
Entrez Gene ID
817748
Gene Name
myosin-like protein XIF
Gene Aliases
AT2G31900, ATMYO5, ATXIF, F20M17.6, F20M17_6, MYOSIN 5, MYOSIN XI F, myosin-like protein XIF
Chromosome Location
Chr.2: 13560370 - 13570469 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336351.1 NP_001318329.1 36 - 37 4974 80
NM_001336352.1 NP_001323600.1 34 - 35 4264 80
NM_001336353.1 NP_001323601.1 29 - 30 3490 80
NM_001336354.1 NP_001323599.1 32 - 33 4055 80
NM_128748.3 NP_180749.2 34 - 35 4261 80

View Full Product Details