Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LUH
Entrez Gene ID
817830
Gene Name
LEUNIG-like protein
Gene Aliases
AT2G32700, F24L7.16, F24L7_16, LEUNIG_homolog, MUCILAGE-MODIFIED 1, MUM1
Chromosome Location
Chr.2: 13866721 - 13872246 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036389.4 NP_001031466.2 10 - 11 1314 83
NM_001202730.1 NP_001189659.1 10 - 11 1359 83
NM_128829.4 NP_565749.1 10 - 11 1314 83
NM_179861.3 NP_850192.1 10 - 11 1260 83
NM_179862.2 NP_850193.1 9 - 10 1389 83
NM_179863.3 NP_850194.1 10 - 11 1356 83
NM_179864.3 NP_850195.2 10 - 11 1205 83
GenBank mRNA AF367306.1 - 10 - 11 1302 83
BT002219.1 - 8 - 9 1035 83

View Full Product Details