Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGO6
Entrez Gene ID
817856
Gene Name
Argonaute family protein
Gene Aliases
ARGONAUTE 6, AT2G32940, T21L14.12, T21L14_12
Chromosome Location
Chr.2: 13971916 - 13977521 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128854.4 NP_180853.2 3 - 4 574 67

View Full Product Details