Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATMRK1
Entrez Gene ID
825501
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT3G63260
Chromosome Location
Chr.3: 23372878 - 23375405 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116191.3 NP_191885.1 3 - 4 1315 61
NM_202754.2 NP_974483.1 3 - 4 1248 61
GenBank mRNA AB006810.1 - 3 - 4 916 61
AY080717.1 - 3 - 4 865 61
AY117337.1 - 3 - 4 657 61

View Full Product Details