Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARV2
Entrez Gene ID
826763
Gene Name
Arv1-like protein
Gene Aliases
AT4G01510, F11O4.16, F11O4_16
Chromosome Location
Chr.4: 642714 - 644400 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340322.1 NP_001328722.1 2 - 3 180 136
NM_001340323.1 NP_001328723.1 2 - 3 180 136
NM_001340326.1 NP_001328725.1 2 - 3 161 136
NM_116381.4 NP_192060.2 2 - 3 180 136
GenBank mRNA AY758071.1 - 2 - 3 161 136

View Full Product Details