Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARV2
Entrez Gene ID
826763
Gene Name
Arv1-like protein
Gene Aliases
AT4G01510, F11O4.16, F11O4_16
Chromosome Location
Chr.4: 642714 - 644400 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340323.1 NP_001328723.1 7 - 8 546 149
NM_001340324.1 NP_001328724.1 7 - 8 535 149
NM_001340325.1 NP_001328720.1 6 - 7 623 149
NM_001340327.1 NP_001328721.1 6 - 7 608 149
NM_001340328.1 NP_001328726.1 5 - 6 458 149
NM_001340329.1 NP_001328727.1 3 - 4 420 149
NM_116381.4 NP_192060.2 7 - 8 546 149
GenBank mRNA AY758071.1 - 7 - 8 527 149

View Full Product Details