Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPK21
Entrez Gene ID
825807
Gene Name
calcium-dependent protein kinase 21
Gene Aliases
AT4G04720, AtCPK21, T4B21.13, T4B21_13, calcium-dependent protein kinase 21
Chromosome Location
Chr.4: 2393653 - 2397804 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340500.1 NP_001319867.1 5 - 6 1412 95
NM_116710.3 NP_192381.1 5 - 6 1457 95
GenBank mRNA AY150502.1 - 5 - 6 1284 95

View Full Product Details