Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAPX
Entrez Gene ID
826396
Gene Name
stromal ascorbate peroxidase
Gene Aliases
AT4G08390, STROMAL ASCORBATE PEROXIDASE, T28D5.80, T28D5_80, stromal ascorbate peroxidase
Chromosome Location
Chr.4: 5314871 - 5317963 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084887.2 NP_001078356.2 3 - 4 696 98
NM_001203761.1 NP_001190690.1 3 - 4 699 98
NM_001340582.1 NP_001319883.1 3 - 4 733 98
NM_116908.3 NP_192579.1 3 - 4 711 98
NM_202791.3 NP_974520.1 3 - 4 699 98
GenBank mRNA AY056319.1 - 3 - 4 637 98
AY114065.1 - 3 - 4 605 98

View Full Product Details