Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SULTR1;1
Entrez Gene ID
826426
Gene Name
sulfate transporter 1;1
Gene Aliases
AT4G08620, SULTR1.2, sulphate transporter 1;1
Chromosome Location
Chr.4: 5500313 - 5506313 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116931.3 NP_192602.1 11 - 12 1802 82

View Full Product Details