Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XRN4
Entrez Gene ID
841891
Gene Name
exoribonuclease 4
Gene Aliases
5'-3' EXORIBONUCLEASE, ACC INSENSITIVE 1, AIN1, AT1G54490, ATXRN4, EIN5, ETHYLENE INSENSITIVE 5, EXORIBONUCLEASE 4, F20D21.30, F20D21_30, exoribonuclease 4
Chromosome Location
Chr.1: 20350109 - 20356916 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333652.1 NP_001321327.1 3 - 4 787 79
NM_104327.4 NP_175851.1 3 - 4 582 79
GenBank mRNA AF286718.1 - 3 - 4 501 79
AY064012.1 - 3 - 4 498 79
AY091411.1 - 3 - 4 391 79
BT026022.1 - 3 - 4 477 79

View Full Product Details