Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZIP4
Entrez Gene ID
842113
Gene Name
Clathrin adaptor complexes medium subunit family protein
Gene Aliases
AP3M, AT1G56590, F25P12.96, F25P12_96, ZIG SUPPRESSOR 4, adaptor protein-3 mu-adaptin
Chromosome Location
Chr.1: 21201948 - 21204856 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104536.5 NP_176052.3 5 - 6 736 78

View Full Product Details