Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRR28
Entrez Gene ID
842263
Gene Name
Tetratricopeptide repeat (TPR)-like superfamily protein
Gene Aliases
AT1G59720, CHLORORESPIRATORY REDUCTION28, F23H11.3, F23H11_3
Chromosome Location
Chr.1: 21939752 - 21941784 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104664.2 NP_176180.1 1 - 1 1559 83

View Full Product Details