Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AIR9
Entrez Gene ID
818033
Gene Name
Outer arm dynein light chain 1 protein
Gene Aliases
AT2G34680, AUXIN-INDUCED IN ROOT CULTURES 9, T29F13.11, T29F13_11
Chromosome Location
Chr.2: 14616601 - 14629207 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336523.1 NP_001324697.1 18 - 19 3090 151
NM_129022.8 NP_181015.7 18 - 19 2901 151

View Full Product Details