Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EDA18
Entrez Gene ID
818057
Gene Name
RING/U-box superfamily protein
Gene Aliases
AT2G34920, F19I3.15, F19I3_15, embryo sac development arrest 18
Chromosome Location
Chr.2: 14728027 - 14731255 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336534.1 NP_001324447.1 5 - 6 2323 67
NM_129045.2 NP_181038.2 5 - 6 2182 67

View Full Product Details