Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNL1
Entrez Gene ID
821265
Gene Name
SIN3-like 1
Gene Aliases
AT3G01320, SIN3-like 1, T22N4.5, T22N4_5
Chromosome Location
Chr.3: 106294 - 113679 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202860.2 NP_001189789.1 16 - 17 3734 133
NM_110998.5 NP_186781.4 16 - 17 3774 133

View Full Product Details