Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPDK
Entrez Gene ID
827226
Gene Name
pyruvate orthophosphate dikinase
Gene Aliases
AT4G15530, DL3805C, FCAALL.325, pyruvate orthophosphate dikinase
Chromosome Location
Chr.4: 8864615 - 8870979 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036570.1 NP_001031647.1 17 - 18 2808 63
NM_001084926.1 NP_001078395.1 16 - 17 2745 63
NM_001084927.2 NP_001078396.1 15 - 16 2797 63
NM_001125515.1 NP_001118987.1 15 - 16 2690 63
NM_001341051.1 NP_001328869.1 15 - 16 2916 63
NM_117643.4 NP_193288.2 14 - 15 2546 63
NM_179060.4 NP_849391.2 16 - 17 2784 63
GenBank mRNA AK317187.1 - 16 - 17 2773 63
AK317578.1 - 14 - 15 2464 63

View Full Product Details