Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPDK
Entrez Gene ID
827226
Gene Name
pyruvate orthophosphate dikinase
Gene Aliases
AT4G15530, DL3805C, FCAALL.325, pyruvate orthophosphate dikinase
Chromosome Location
Chr.4: 8864615 - 8870979 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036570.1 NP_001031647.1 18 - 19 2985 74
NM_001084926.1 NP_001078395.1 17 - 18 2922 74
NM_001084927.2 NP_001078396.1 16 - 17 2974 74
NM_001125515.1 NP_001118987.1 16 - 17 2867 74
NM_001341051.1 NP_001328869.1 16 - 17 3093 74
NM_117643.4 NP_193288.2 15 - 16 2723 74
NM_179060.4 NP_849391.2 17 - 18 2961 74
GenBank mRNA AK317187.1 - 17 - 18 2950 74
AK317578.1 - 15 - 16 2641 74

View Full Product Details