Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCG39
- Entrez Gene ID
-
843013
- Gene Name
- pleiotropic drug resistance 11
- Gene Aliases
-
AT1G66950, ATP-binding cassette G39, ATPDR11, PDR11, PLEIOTROPIC DRUG RESISTANCE 11, T4O24.9, T4O24_9, pleiotropic drug resistance 11
- Chromosome Location
-
Chr.1: 24978121 - 24984619 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105366.4 |
NP_176867.2 |
1 - 2 |
505 |
169 |
View Full Product Details