Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SERK1
Entrez Gene ID
843513
Gene Name
somatic embryogenesis receptor-like kinase 1
Gene Aliases
AT1G71830, ATSERK1, F14O23.21, F14O23_21, SOMATIC EMBRYOGENESIS RECEPTOR-LIKE KINASE 1, somatic embryogenesis receptor-like kinase 1
Chromosome Location
Chr.1: 27018158 - 27022117 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105841.4 NP_177328.1 1 - 2 504 90
GenBank mRNA FJ708676.1 - 1 - 2 87 90

View Full Product Details