Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAS1
Entrez Gene ID
823649
Gene Name
lanosterol synthase 1
Gene Aliases
AT3G45130, lanosterol synthase 1
Chromosome Location
Chr.3: 16512170 - 16517781 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339194.1 NP_001327790.1 2 - 3 340 133
NM_001339195.1 NP_001327789.1 1 - 2 247 133
NM_001339196.1 NP_001327788.1 1 - 2 447 133
NM_001339197.1 NP_001327787.1 2 - 3 340 133
NM_114382.5 NP_190099.3 1 - 2 228 133
GenBank mRNA AB247155.1 - 1 - 2 188 133
DQ508794.2 - 1 - 2 188 133

View Full Product Details