Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FUS5
Entrez Gene ID
839241
Gene Name
Proteasome component (PCI) domain protein
Gene Aliases
ARABIDOPSIS THALIANA COP9 SIGNALOSOME SUBUNIT 7, AT1G02090, ATCSN7, CONSTITUTIVE PHOTOMORPHOGENIC 15, COP15, COP9 SIGNALOSOME SUBUNIT 7, CSN7, FUSCA 5, T7I23.25, T7I23_25
Chromosome Location
Chr.1: 387277 - 389808 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100089.3 NP_563645.1 8 - 9 999 77
NM_179245.3 NP_849576.1 8 - 9 982 77
NM_202010.3 NP_973739.1 7 - 8 1594 77
GenBank mRNA AF063852.2 - 8 - 9 786 77
AF372934.1 - 8 - 9 834 77
AY087971.1 - 8 - 9 837 77

View Full Product Details