Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HST
Entrez Gene ID
819666
Gene Name
ARM repeat superfamily protein
Gene Aliases
AT3G05040, HASTY, HASTY 1, HST1, T9J14.1, T9J14_1
Chromosome Location
Chr.3: 1401171 - 1408302 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337559.1 NP_001327303.1 18 - 19 2742 93
NM_111376.4 NP_187155.2 18 - 19 2754 93
GenBank mRNA AK229239.1 - 18 - 19 2649 93

View Full Product Details