Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBP31
Entrez Gene ID
828579
Gene Name
31-kDa RNA binding protein
Gene Aliases
31-kDa RNA binding protein, APPROXIMATELY 31 KD, ARABIDOPSIS THALIANA RNA BINDING PROTEIN, AT4G24770, ATRBP31, ATRBP33, CP31, F6I7.11, STEP1
Chromosome Location
Chr.4: 12765761 - 12769419 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341706.1 NP_001328008.1 2 - 3 745 63
NM_118610.4 NP_194208.1 1 - 2 826 63
GenBank mRNA M94554.1 - 1 - 2 578 63
X65255.1 - - 590 63

View Full Product Details