Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl17
Entrez Gene ID
820399
Gene Name
serine carboxypeptidase-like 17
Gene Aliases
AT3G12203, serine carboxypeptidase-like 17
Chromosome Location
Chr.3: 3891160 - 3894631 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337980.1 NP_001327414.1 11 - 12 1016 139
NM_001337981.1 NP_001327415.1 11 - 12 1022 139
NM_112056.3 NP_187828.1 10 - 11 982 139
GenBank mRNA BT012565.1 - 10 - 11 957 139

View Full Product Details