Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PME3
Entrez Gene ID
820651
Gene Name
pectin methylesterase 3
Gene Aliases
AT3G14310, ATPME3, OVERLY ZINC SENSITIVE 2, OZS2, pectin methylesterase 3
Chromosome Location
Chr.3: 4771833 - 4775258 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112289.3 NP_188048.1 1 - 2 761 70
GenBank mRNA AY037184.1 - 1 - 2 622 70
AY052252.1 - 1 - 2 619 70
AY143950.1 - 1 - 2 598 70

View Full Product Details