Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LBD13
Entrez Gene ID
817584
Gene Name
LOB domain-containing protein 13
Gene Aliases
AT2G30340, LOB domain-containing protein 13, T9D9.15, T9D9_15
Chromosome Location
Chr.2: 12931172 - 12932828 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336251.1 NP_001323878.1 1 - 2 672 62
NM_179810.3 NP_850141.1 2 - 3 582 62
GenBank mRNA AB473843.1 - 2 - 3 302 62
AF447888.1 - 2 - 3 336 62
BT026052.1 - 1 - 2 294 62

View Full Product Details