Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COBL8
Entrez Gene ID
820941
Gene Name
COBRA-like protein 8 precursor
Gene Aliases
AT3G16860, COBRA-like protein 8 precursor
Chromosome Location
Chr.3: 5759375 - 5762264 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112562.4 NP_188311.1 1 - 2 700 100

View Full Product Details