Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CIPK1
Entrez Gene ID
821016
Gene Name
CBL-interacting protein kinase 1
Gene Aliases
AT3G17510, CBL-interacting protein kinase 1, SNF1-RELATED PROTEIN KINASE 3.16, SnRK3.16
Chromosome Location
Chr.3: 5988924 - 5993001 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112631.3 NP_566580.1 7 - 8 1285 81
NM_202599.2 NP_974328.1 5 - 6 1534 81
GenBank mRNA BT029495.1 - 5 - 6 671 81

View Full Product Details