Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IPT8
Entrez Gene ID
821450
Gene Name
ATP/ADP isopentenyltransferase
Gene Aliases
AT3G19160, ATIPT8, ATP/ADP isopentenyltransferases, MVI11.16, PGA22, isopentenyltransferase 8
Chromosome Location
Chr.3: 6621988 - 6623278 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112803.1 NP_188547.1 1 - 2 557 82
GenBank mRNA AB061406.1 - 1 - 2 557 82
AB062614.1 - 1 - 2 515 82

View Full Product Details