Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APX5
Entrez Gene ID
829751
Gene Name
ascorbate peroxidase 5
Gene Aliases
AT4G35970, T19K4.100, ascorbate peroxidase 5
Chromosome Location
Chr.4: 17028586 - 17030558 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342391.1 NP_001328787.1 7 - 8 603 66
NM_119763.3 NP_195321.1 7 - 8 623 66
GenBank mRNA AK119023.1 - 7 - 8 603 66
BT006053.1 - 7 - 8 558 66

View Full Product Details