Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP81F3
- Entrez Gene ID
-
829894
- Gene Name
- cytochrome P450, family 81, subfamily F, polypeptide 3
- Gene Aliases
-
''cytochrome P450, AT4G37400, F6G17.50, F6G17_50, cytochrome P450, family 81, polypeptide 3, polypeptide 3'', subfamily F
- Chromosome Location
-
Chr.4: 17583992 - 17586442 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119903.4 |
NP_568025.1 |
1 - 2 |
973 |
82 |
GenBank mRNA |
AK227603.1 |
- |
1 - 2 |
914 |
82 |
View Full Product Details