Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAC3
Entrez Gene ID
818382
Gene Name
acetyl Co-enzyme a carboxylase carboxyltransferase alpha subunit
Gene Aliases
AT2G38040, CARBOXYLTRANSFERASE ALPHA SUBUNIT, T8P21.5, T8P21_5, acetyl Co-enzyme a carboxylase carboxyltransferase alpha subunit
Chromosome Location
Chr.2: 15917032 - 15921170 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179960.1 NP_850291.1 11 - 12 2626 95

View Full Product Details