Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AO4
- Entrez Gene ID
-
839488
- Gene Name
- aldehyde oxidase 4
- Gene Aliases
-
AAO4, ALDEHYDE OXIDASE 2, ALDEHYDE OXIDASE 4, AT1G04580, ATAO-4, ATAO2, T1G11.17, T1G11_17, aldehyde oxidase 4, arabdopsis thaliana aldehyde oxidase 4
- Chromosome Location
-
Chr.1: 1252005 - 1257893 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100337.3 |
NP_563711.1 |
7 - 8 |
3876 |
100 |
GenBank mRNA |
AB037271.1 |
- |
7 - 8 |
3493 |
100 |
View Full Product Details