Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PKT4
Entrez Gene ID
839434
Gene Name
peroxisomal 3-ketoacyl-CoA thiolase 4
Gene Aliases
3-KETO-ACYL-COA THIOLASE 1, AT1G04710, KAT1, T1G11.4, T1G11_4, peroxisomal 3-ketoacyl-CoA thiolase 4, peroxisomal 3-ketoacyl-CoA thiolase 4
Chromosome Location
Chr.1: 1321885 - 1324840 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100351.5 NP_171965.1 13 - 14 1314 70
GenBank mRNA AY099741.1 - 13 - 14 1290 70
BT003415.1 - 13 - 14 1258 70

View Full Product Details