Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ROPGEF4
- Entrez Gene ID
-
819197
- Gene Name
- RHO guanyl-nucleotide exchange factor 4
- Gene Aliases
-
AT2G45890, ATROPGEF4, F4I18.13, GEF4, RHO GUANYL-NUCLEOTIDE EXCHANGE FACTOR 4, RHO guanyl-nucleotide exchange factor 4, RHS11, ROOT HAIR SPECIFIC 11, ROP (rho of plants) guanine nucleotide exchange factor 4, guanine exchange factor 4
- Chromosome Location
-
Chr.2: 18883377 - 18885729 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_130152.3 |
NP_182113.2 |
5 - 6 |
1088 |
78 |
GenBank mRNA |
AK229390.1 |
- |
5 - 6 |
992 |
78 |
View Full Product Details