Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HA3
Entrez Gene ID
835841
Gene Name
H[+]-ATPase 3
Gene Aliases
AHA3, ARABIDOPSIS THALIANA ARABIDOPSIS H(+)-ATPASE, AT5G57350, ATAHA3, H(+)-ATPase 3, MJB24.16, MJB24_16, P-TYPE H(+)-ATPASE ISOFORM 3
Chromosome Location
Chr.5: 23230874 - 23236682 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203630.1 NP_001190559.1 7 - 8 1424 78
NM_001345258.1 NP_001331203.1 3 - 4 1203 78
NM_001345259.1 NP_001331204.1 7 - 8 1478 78
NM_125118.3 NP_200545.1 7 - 8 1540 78
GenBank mRNA AY072153.1 - 7 - 8 1420 78

View Full Product Details